Learn More
Medchemexpress LLC HY-12750 50mg , AZD3965 CAS:1448671-31-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1275050MG
Encompass_Preferred
HY-12750 50mg AZD3965 CAS: 1448671-31-5 AZD3965 is a selective MCT1 inhibitor with a Ki of 1.6 nM, showing 6-fold selectivity over MCT2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0012