Learn More
Medchemexpress LLC HY-50861 5mg Medchemexpress, AZD3988 CAS:892489-52-0 Purity:>98%
SDP
Catalog No. 501970073
Encompass_Preferred
Medchemexpress, HY-50861 5mg AZD3988 CAS:892489-52-0 AZD3988 is a diacylglycerol acyl transferase-1 (DGAT-1) inhibitor with IC50s of 6, 5, 11 nM for human, rat, mouse, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0073 Supplier Medchemexpress LLC Supplier No. HY508615MG
Medchemexpress, HY-50861 5mg AZD3988 CAS:892489-52-0 AZD3988 is a diacylglycerol acyl transferase-1 (DGAT-1) inhibitor with IC50s of 6, 5, 11 nM for human, rat, mouse, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.