Learn More
Medchemexpress LLC HY-112416 5mg Medchemexpress, AZD4320 CAS:1357576-48-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1124165MG
Encompass_Preferred
Medchemexpress, HY-112416 5mg AZD4320 CAS:1357576-48-7 AZD4320 is a novel BH3-mimicking dual BCL2/BCLxL inhibitor with IC50s of 26 nM, 17 nM, and 170 nM for KPUM-MS3, KPUM-UH1, and STR-428 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9726