Learn More
Selleck Chemical LLC AZD4547 10mg 1035270-39-3
SDP
Catalog No. NC0660421
Encompass_Preferred
AZD4547 is a novel selective FGFR inhibitor targeting FGFR1/2/3 with IC50 of 0.2 nM/2.5 nM/1.8 nM in cell-free assays, weaker activity against FGFR4, VEGFR2(KDR), and little activity observed against IGFR, CDK2, and p38. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0660421 Supplier Selleck Chemical LLC Supplier No. S280110MG
AZD4547 is a novel selective FGFR inhibitor targeting FGFR1/2/3 with IC50 of 0.2 nM/2.5 nM/1.8 nM in cell-free assays, weaker activity against FGFR4, VEGFR2(KDR), and little activity observed against IGFR, CDK2, and p38. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.