Learn More
Medchemexpress LLC HY-13330 10mg Medchemexpress, AZD4547 CAS:1035270-39-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1333010MG
Encompass_Preferred
Medchemexpress, HY-13330 10mg AZD4547 CAS:1035270-39-3 AZD4547 is a potent inhibitor of the FGFR family with IC50s of 0.2 nM, 2.5 nM, 1.8 nM, and 165 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8986
 Restricted Item