Learn More
Medchemexpress LLC HY-112088 10mg , AZD4573 CAS:2057509-72-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11208810MG
Encompass_Preferred
HY-112088 10mg AZD4573 CAS: 2057509-72-3 AZD4573 is a potent and highly selective CDK9 inhibitor (IC50 of <4 nM) that enables transient target engagement for the treatment of hematologic malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9055