Learn More
Selleck Chemical LLC AZD4573 2mg 2057509-72-3
SDP
Supplier:  Selleck Chemical LLC S87192MG
Encompass_Preferred
AZD4573 is a potent inhibitor of CDK9 (IC50 of <0.004 ?M) with fast-off binding kinetics (t1/2 = 16 min) and high selectivity versus other kinases, including other CDK family kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8217