Learn More
Medchemexpress LLC AZD4694 Precursor | 1211333-20-8 | 98.8% | 443.45 | 10 MM 1 ML
SDP
Catalog No. 5000348938
Encompass_Preferred
AZD4694 Precursor (AZ13040214) is the precursor of [18F] AZD4694 for the synthesis of [18F] AZD4694, an amyloid-β imaging ligand with high affinity for amyloid-β plaques.
  • Precursor for the synthesis of [18F] AZD4694.
  • [18F] AZD4694 is an amyloid-β imaging ligand.
  • [18F] AZD4694 exhibits high affinity for amyloid-β plaques.
  • Also known as AZ13040214 and NAV4614.
Catalog No. 50-003-48938 Supplier Medchemexpress LLC Supplier No. HY13951610MM/1ML
AZD4694 Precursor (AZ13040214) is the precursor of [18F] AZD4694 for the synthesis of [18F] AZD4694, an amyloid-β imaging ligand with high affinity for amyloid-β plaques.
  • Precursor for the synthesis of [18F] AZD4694.
  • [18F] AZD4694 is an amyloid-β imaging ligand.
  • [18F] AZD4694 exhibits high affinity for amyloid-β plaques.
  • Also known as AZ13040214 and NAV4614.