Learn More
Selleck Chemical LLC AZD5069-5mg
SDP
Supplier:  Selleck Chemical LLC S66455MG
Encompass_Preferred
AZD5069 is a novel antagonist of CXCR2, which is shown to inhibit binding of CXCL8 to CXCR2 with a pIC50 value of 8.8 and inhibit CXCL8 binding to CXCR1 with pIC50 values of 6.5.
Catalog No. 50-238-9838