Learn More
Selleck Chemical LLC AZD-5153 6-hydroxy-2-naphthoic acid 5mg 1869912-40-2 AZD-5153 HNT salt
SDP
Supplier:  Selleck Chemical LLC S83445MG
Encompass_Preferred
AZD-5153 6-hydroxy-2-naphthoic acid (HNT salt) is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor with pKi of 8.3 for BRD4. AZD-5153 inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes. NSD3, via H3K36me2, acts as an epigenetic deregulator to facilitate the expression of oncogenesis-promoting genes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5971