Learn More
Medchemexpress LLC HY-100653A 100mg , AZD5153 (6-Hydroxy-2-naphthoic acid) AZD-5153 HNT salt CAS:1869912-40-2 Purity:98%
SDP
Catalog No. 501493340
Encompass_Preferred
Medchemexpress, HY-100653A 100mg AZD5153 (6-Hydroxy-2-naphthoic acid) AZD-5153 HNT salt CAS:1869912-40-2 Formula:C36H41N7O6 IC50: 1.7 nM (BRD4) Purity:98% AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor; disrupts BRD4 with an IC 50 of 1.7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3340 Supplier Medchemexpress LLC Supplier No. HY100653A100MG
Medchemexpress, HY-100653A 100mg AZD5153 (6-Hydroxy-2-naphthoic acid) AZD-5153 HNT salt CAS:1869912-40-2 Formula:C36H41N7O6 IC50: 1.7 nM (BRD4) Purity:98% AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor; disrupts BRD4 with an IC 50 of 1.7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.