Learn More
Apexbio Technology LLC AZD5363, 100mg. Cas: 1143532-39-1 MFCD: MFCD22628785. AKT inhibitor,pyrrolopyrimidine derived
SDP
Supplier:  Apexbio Technology LLC A1387100
Encompass_Preferred
AZD5363 is a novel, potent phosphoinositide 3-kinase (PI3K)/Akt pathway inhibitor with IC50 value of ~200nM. [1]AZD5363 is proved to inhibit castrate resistant prostate cancer (CRPC) progression. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4695