Learn More
Medchemexpress LLC HY-15431 100mg , AZD5363 Capivasertib CAS:1143532-39-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15431100MG
Encompass_Preferred
Medchemexpress, HY-15431 100mg AZD5363 Capivasertib CAS:1143532-39-1 Formula:C21H25ClN6O2 IC50: 3 nM (Akt1), 7 nM (Akt2), 7 nM (Akt3), 6 nM (p70S6K), 7 nM (PKA), 60 nM (ROCK2) Purity:98% AZD5363 is a potent pan-AKT kinase inhibitor with IC 50 of 3, 7 and 7 nM for Akt1 , Akt2 and Akt3 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2823