Learn More
Medchemexpress LLC AZD5423 100mg | 1034148-04-3 | 100MG
SDP
Catalog No. 5000242720
Encompass_Preferred
AZD5423 is an inhaled potent selective and non-steroidal glucocorticoid receptor (GR) modulator (SGRM)[1 AZD5423 effectively reduces allergen-induced responses in subjects with mild allergic asthma[2
Catalog No. 50-002-42720 Supplier Medchemexpress LLC Supplier No. HY108243100MG
AZD5423 is an inhaled potent selective and non-steroidal glucocorticoid receptor (GR) modulator (SGRM)[1 AZD5423 effectively reduces allergen-induced responses in subjects with mild allergic asthma[2