Learn More
Medchemexpress LLC HY-108243 10mg Medchemexpress, AZD5423 CAS:1034148-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10824310MG
Encompass_Preferred
Medchemexpress, HY-108243 10mg AZD5423 CAS:1034148-04-3 AZD5423 is an inhaled, potent, selective, and non-steroidal glucocorticoid receptor (GR) modulator (SGRM). AZD5423 effectively reduces allergen-induced responses in subjects with mild allergic asthma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8512