Learn More
Selleck Chemical LLC AZD5582 5mg 1258392-53-8
SDP
Supplier:  Selleck Chemical LLC S73625MG
Encompass_Preferred
AZD5582, a novel small-molecule IAP inhibitor, binds potently to the BIR3 domains of cIAP1, cIAP2, and XIAP with IC50 values of 15, 21, and 15 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4687