Learn More
Selleck Chemical LLC AZD5991-5mg
SDP
Supplier:  Selleck Chemical LLC S86435MG
Encompass_Preferred
AZD5991 is a macrocyclic MCL-1 inhibitor with sub-nanomolar affinity for MCL-1 (Ki = 0.13 nM). The binding affinity of AZD5991 is about 25-fold lower for mouse Mcl-1 vs. human Mcl-1 but only four-fold lower for rat Mcl-1.
Catalog No. 50-238-9790