Learn More
Abcam AZD6244, MEK1/2 inhibitor, 25MG
Catalog No. 502706670
Encompass_Preferred
MW 457.7 Da, Purity >99%. Potent, selective, non-ATP competitive MEK1/2 inhibitor (IC₅₀ MEK1 = 14 nM). Also inhibits ERK1/2 phosphorylation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6670 Supplier Abcam Supplier No. AB14134325MG
MW 457.7 Da, Purity >99%. Potent, selective, non-ATP competitive MEK1/2 inhibitor (IC₅₀ MEK1 = 14 nM). Also inhibits ERK1/2 phosphorylation.
The product is subject to the following: Abcam Restricted Use Statement