Learn More
Abcam AZD6244, MEK1/2 inhibitor, 5MG
Catalog No. 502706669
Encompass_Preferred
MW 457.7 Da, Purity >99%. Potent, selective, non-ATP competitive MEK1/2 inhibitor (IC₅₀ MEK1 = 14 nM). Also inhibits ERK1/2 phosphorylation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6669 Supplier Abcam Supplier No. AB1413435MG
MW 457.7 Da, Purity >99%. Potent, selective, non-ATP competitive MEK1/2 inhibitor (IC₅₀ MEK1 = 14 nM). Also inhibits ERK1/2 phosphorylation.
The product is subject to the following: Abcam Restricted Use Statement