Learn More
Apexbio Technology LLC AZD6244 (Selumetinib), 606143-52-6, MFCD11977472, 100 mg
SDP
Supplier:  Apexbio Technology LLC A8207100
Encompass_Preferred
MF: C17H15BrClFN4O3, MW: 457.69, Solubility: 22.9mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. AZD6244 is a highly potent and selective inhibitor of MEK1/2 with IC50 value of 14.1nM against MEK1.
Catalog No. 50-101-3727