Learn More
Selleck Chemical LLC AZD6482 50mg 1173900-33-8 KIN-193
SDP
Catalog No. 508673
Encompass_Preferred
AZD6482 (KIN-193) is a PI3K? inhibitor with IC50 of 10 nM, 8-, 87- and 109-fold more selective to PI3K? than PI3K?, PI3K? and PI3K? in cell-free assays. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-867-3 Supplier Selleck Chemical LLC Supplier No. S146250MG
AZD6482 (KIN-193) is a PI3K? inhibitor with IC50 of 10 nM, 8-, 87- and 109-fold more selective to PI3K? than PI3K?, PI3K? and PI3K? in cell-free assays. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.