Learn More
Selleck Chemical LLC Ceralasertib (AZD6738) 5mg 1352226-88-0
SDP
Catalog No. NC1029631
Encompass_Preferred
Ceralasertib (AZD6738) is an orally active, and selective ATR kinase inhibitor with IC50 of 1 nM. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1029631 Supplier Selleck Chemical LLC Supplier No. S76935MG
Ceralasertib (AZD6738) is an orally active, and selective ATR kinase inhibitor with IC50 of 1 nM. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.