Learn More
Apexbio Technology LLC AZD6738, 1352226-88-0, MFCD28952790, 5 mg
SDP
Supplier:  Apexbio Technology LLC B60075
Encompass_Preferred
MF: C20H24N6O2S, MW: 412.51, Solubility: 82 mg/mL in DMSO, 82.4 mg/mL in EtOH, insoluble in H2O. AZD6738 is a selective ATR (Ataxia Telangiectasia and RAD3-related) inhibitor, with an IC50 of approximately 30 uM to resting CLL cells.
Catalog No. 50-115-2496