Learn More
Medchemexpress LLC HY-111453 5mg Medchemexpress, AZD7594 CAS:1196509-60-0 Purity:>98%
SDP
Catalog No. 501968872
Encompass_Preferred
Medchemexpress, HY-111453 5mg AZD7594 CAS:1196509-60-0 AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8872 Supplier Medchemexpress LLC Supplier No. HY1114535MG
Medchemexpress, HY-111453 5mg AZD7594 CAS:1196509-60-0 AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.