Learn More
Selleck Chemical LLC AZD7648 5mg
SDP
Supplier:  Selleck Chemical LLC S88435MG
Encompass_Preferred
AZD7648 is a potent inhibitor of DNA-PK with an IC50 of 0.6 nM in biochemical assay and more than 100-fold selective against 396 other kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3399