Learn More
Apexbio Technology LLC AZD7762(Synonyms: AZD-7762, AZD 7762, 860352-01-8), 10mM (in 1mL DMSO), CAS: 860352-01-8.
SDP
Catalog No. 5000060683
Encompass_Preferred
AZD7762 (CAS No 860352-01-8) is an ATP-competitive inhibitor targeting the checkpoint kinases Chk1 and Chk2 regulatory enzymes activated upon DNA damage that control cell-cycle progression through phosphorylation of CDC25 phosphatases By competitively binding the ATP-binding pocket of Chk1 AZD7762 blocks Chk1-mediated phosphorylation of CDC25C exhibiting an IC50 of approximately 5 nM and Ki of 3 6 nM This inhibition prevents DNA damage-induced cell cycle arrest and interrupts DNA repair mechanisms promoting apoptosis in tumor cells Consequently AZD7762 acts as a chemosensitizing agent enhancing the cytotoxic effects of DNA-damaging therapies in oncology research
Catalog No. 50-000-60683 Supplier Apexbio Technology LLC Supplier No. A59195.1
AZD7762 (CAS No 860352-01-8) is an ATP-competitive inhibitor targeting the checkpoint kinases Chk1 and Chk2 regulatory enzymes activated upon DNA damage that control cell-cycle progression through phosphorylation of CDC25 phosphatases By competitively binding the ATP-binding pocket of Chk1 AZD7762 blocks Chk1-mediated phosphorylation of CDC25C exhibiting an IC50 of approximately 5 nM and Ki of 3 6 nM This inhibition prevents DNA damage-induced cell cycle arrest and interrupts DNA repair mechanisms promoting apoptosis in tumor cells Consequently AZD7762 acts as a chemosensitizing agent enhancing the cytotoxic effects of DNA-damaging therapies in oncology research