Learn More
Selleck Chemical LLC AZD7762 S1532-2mg
SDP
Catalog No. 503108335
Encompass_Preferred
AZD7762 is a potent and selective inhibitor of Chk1 with IC50 of 5 nM in a cell-free assay It is equally potent against Chk2 and less potent against CAM Yes Fyn Lyn Hck and Lck Phase 1
Catalog No. 50-310-8335 Supplier Selleck Chemical LLC Supplier No. S15322MG
AZD7762 is a potent and selective inhibitor of Chk1 with IC50 of 5 nM in a cell-free assay It is equally potent against Chk2 and less potent against CAM Yes Fyn Lyn Hck and Lck Phase 1