Learn More
Selleck Chemical LLC AZD7762 5mg 860352-01-8
SDP
Catalog No. 508038
Encompass_Preferred
AZD7762 is a potent and selective inhibitor of Chk1 with IC50 of 5 nM in a cell-free assay. It is equally potent against Chk2 and less potent against CAM, Yes, Fyn, Lyn, Hck and Lck. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-803-8 Supplier Selleck Chemical LLC Supplier No. S15325MG
AZD7762 is a potent and selective inhibitor of Chk1 with IC50 of 5 nM in a cell-free assay. It is equally potent against Chk2 and less potent against CAM, Yes, Fyn, Lyn, Hck and Lck. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.