Learn More
Medchemexpress LLC HY-101056 5mg Medchemexpress, AZD7986 CAS:1802148-05-5 Purity:>98%
SDP
Catalog No. 501932024
Encompass_Preferred
Medchemexpress, HY-101056 5mg AZD7986 CAS:1802148-05-5 AZD7986 is a Dipeptidyl peptidase 1 ( DPP1 ) inhibitor with pIC 50 s of 6.85, 7.6, 7.7, 7.8, and 7.8 in human, mouse, rat, dog and rabbit, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2024 Supplier Medchemexpress LLC Supplier No. HY1010565MG
Medchemexpress, HY-101056 5mg AZD7986 CAS:1802148-05-5 AZD7986 is a Dipeptidyl peptidase 1 ( DPP1 ) inhibitor with pIC 50 s of 6.85, 7.6, 7.7, 7.8, and 7.8 in human, mouse, rat, dog and rabbit, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.