Learn More
Selleck Chemical LLC AZD8055 10mg 1009298-09-2
SDP
Supplier:  Selleck Chemical LLC S155510MG
Encompass_Preferred
AZD8055 is a novel ATP-competitive mTOR inhibitor with IC50 of 0.8 nM in MDA-MB-468 cells with excellent selectivity (?1,000-fold) against PI3K isoforms and ATM/DNA-PK. AZD8055 induces caspase-dependent apoptosis and also induces autophagy. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-868-1