Learn More
Medchemexpress LLC HY-12330 10mg , AZD8186 CAS:1627494-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1233010MG
Encompass_Preferred
HY-12330 10mg AZD8186 CAS: 1627494-13-6 AZD8186 is a PI3K inhibitor, which potently inhibits PI3Kβ (IC50=4 nM) and PI3Kδ (IC5050=12 nM) with selectivity over PI3Kα (IC50=35 nM) and PI3Kγ (IC50=675 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9658