Learn More
Medchemexpress LLC HY-18173 5mg Medchemexpress, AZD8329 CAS:1048668-70-7 Purity:>98%
SDP
Catalog No. 501932808
Encompass_Preferred
Medchemexpress, HY-18173 5mg AZD8329 CAS:1048668-70-7 AZD8329 is a potent 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC 50 of 9 nM for human 11β-HSD1, displays excellent selectivity versus 11β-HSD2, 17β-HSD1 and 17β-HSD3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2808 Supplier Medchemexpress LLC Supplier No. HY181735MG
Medchemexpress, HY-18173 5mg AZD8329 CAS:1048668-70-7 AZD8329 is a potent 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC 50 of 9 nM for human 11β-HSD1, displays excellent selectivity versus 11β-HSD2, 17β-HSD1 and 17β-HSD3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.