Learn More
Selleck Chemical LLC AZD8330 5mg 869357-68-6 ARRY704
SDP
Catalog No. 501362097
Encompass_Preferred
AZD8330 (ARRY704) is a novel, selective, non-ATP competitive MEK 1/2 inhibitor with IC50 of 7 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2097 Supplier Selleck Chemical LLC Supplier No. S21345MG
AZD8330 (ARRY704) is a novel, selective, non-ATP competitive MEK 1/2 inhibitor with IC50 of 7 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.