Learn More
Medchemexpress LLC HY-19427A 100mg , AZD9056 (hydrochloride) CAS:345303-91-5 Purity:98%
SDP
Catalog No. 501493208
Encompass_Preferred
Medchemexpress, HY-19427A 100mg AZD9056 (hydrochloride) CAS:345303-91-5 Formula:C24H36Cl2N2O2 Purity:98% AZD9056 is a selective orally active inhibitor of P2X7 which plays a significant role in inflammation and pain-causing diseases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3208 Supplier Medchemexpress LLC Supplier No. HY19427A100MG
Medchemexpress, HY-19427A 100mg AZD9056 (hydrochloride) CAS:345303-91-5 Formula:C24H36Cl2N2O2 Purity:98% AZD9056 is a selective orally active inhibitor of P2X7 which plays a significant role in inflammation and pain-causing diseases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.