Learn More
Selleck Chemical LLC Osimertinib (AZD9291) 5mg 1421373-65-0
SDP
Catalog No. NC0845425
Encompass_Preferred
Osimertinib (AZD9291) is an oral, irreversible, and mutant-selective EGFR inhibitor with IC50 of 12.92, 11.44 and 493.8 nM for Exon 19 deletion EGFR, L858R/T790M EGFR, and WT EGFR in LoVo cells, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0845425 Supplier Selleck Chemical LLC Supplier No. S72975MG
Osimertinib (AZD9291) is an oral, irreversible, and mutant-selective EGFR inhibitor with IC50 of 12.92, 11.44 and 493.8 nM for Exon 19 deletion EGFR, L858R/T790M EGFR, and WT EGFR in LoVo cells, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.