Learn More
Selleck Chemical LLC AZD9496
SDP
Catalog No. 5000002877
Encompass_Preferred
AZD9496 is an oral estrogen receptor inhibitor that blocks the growth of ER-positive and ESR1 mutant breast tumours in preclinical models
Catalog No. 50-000-02877 Supplier Selleck Chemical LLC Supplier No. S83722MG
AZD9496 is an oral estrogen receptor inhibitor that blocks the growth of ER-positive and ESR1 mutant breast tumours in preclinical models