Learn More
TARGETMOL CHEMICALS INC AZD9496 5MG
SDP
Catalog No. 502576849
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZD9496 is an orally available selective estrogen receptor(ER(alpha)) antagonist with potential antineoplastic activity. purity: 98%
Catalog No. 50-257-6849 Supplier TARGETMOL CHEMICALS INC Supplier No. T39755MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZD9496 is an orally available selective estrogen receptor(ER(alpha)) antagonist with potential antineoplastic activity. purity: 98%