Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AZD9496 deacrylic acid phenol | 2173404-70-9 | 96.6% | C22H23F3N2O | 5 MG
SDP

Catalog No. 5000359274
Encompass_Preferred

AZD9496 deacrylic acid phenol is a fragment of a target protein ligand of PROTAC ER Degrader-4. This product is for research use only.

  • Purity: 96.6%
  • Appearance: Light yellow to yellow solid
  • Storage (solid form): -20°C, stored under nitrogen
  • Shipping: Room temperature

Catalog No. 50-003-59274 Supplier Medchemexpress LLC Supplier No. HY463405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZD9496 deacrylic acid phenol is a fragment of a target protein ligand of PROTAC ER Degrader-4. This product is for research use only.

  • Purity: 96.6%
  • Appearance: Light yellow to yellow solid
  • Storage (solid form): -20°C, stored under nitrogen
  • Shipping: Room temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.