Learn More
Medchemexpress LLC AZD9496 maleate | 1639042-28-6 | 99.4% | 50 MG
SDP
Catalog No. 5000341019
Encompass_Preferred
AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with an IC50 of 0.28 nM. It functions as an orally bioavailable selective estrogen receptor degrader (SERD), identified as a laboratory chemical.
  • Potent and selective estrogen receptor (ERα) antagonist
  • Orally bioavailable selective estrogen receptor degrader (SERD)
  • Suitable for scientific research
  • Used in signaling pathways
  • For research use only
Catalog No. 50-003-41019 Supplier Medchemexpress LLC Supplier No. HY12870A50MG
AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with an IC50 of 0.28 nM. It functions as an orally bioavailable selective estrogen receptor degrader (SERD), identified as a laboratory chemical.
  • Potent and selective estrogen receptor (ERα) antagonist
  • Orally bioavailable selective estrogen receptor degrader (SERD)
  • Suitable for scientific research
  • Used in signaling pathways
  • For research use only