Learn More
Medchemexpress LLC HY-12870A 5mg Medchemexpress, AZD9496 (maleate) CAS:1639042-28-6 Purity:>98%
SDP
Catalog No. 502081176
Encompass_Preferred
Medchemexpress, HY-12870A 5mg AZD9496 (maleate) CAS:1639042-28-6 AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with IC50 of 0.28 nM. AZD9496 maleate is an orally bioavailable selective oestrogen receptor degrader (SERD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1176 Supplier Medchemexpress LLC Supplier No. HY12870A5MG
Medchemexpress, HY-12870A 5mg AZD9496 (maleate) CAS:1639042-28-6 AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with IC50 of 0.28 nM. AZD9496 maleate is an orally bioavailable selective oestrogen receptor degrader (SERD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.