Learn More
Medchemexpress LLC AZD9496 maleate | ≥95.0% | 2 MG
SDP
Catalog No. 5000341921
Encompass_Preferred
AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist. It acts as an orally bioavailable selective oestrogen receptor degrader (SERD).
  • Potent and selective ERα antagonist
  • Orally bioavailable selective oestrogen receptor degrader (SERD)
  • IC50 of 0.28 nM
Catalog No. 50-003-41921 Supplier Medchemexpress LLC Supplier No. HY12870A2MG
AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist. It acts as an orally bioavailable selective oestrogen receptor degrader (SERD).
  • Potent and selective ERα antagonist
  • Orally bioavailable selective oestrogen receptor degrader (SERD)
  • IC50 of 0.28 nM