Learn More
Medchemexpress LLC HY-120012 10mg Medchemexpress, AZD9567 CAS:1893415-00-3 Purity:>98%
SDP
Catalog No. 501969567
Encompass_Preferred
Medchemexpress, HY-120012 10mg AZD9567 CAS:1893415-00-3 AZD9567 (compound 15) is a potent, oral active, non-steroidal and selective glucocorticoid receptor modulator (SGRM), with an IC50 of 3.8 nM. Exhibits excellent efficacy in the streptococcal cell wall (SCW) reactivation model of joint inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9567 Supplier Medchemexpress LLC Supplier No. HY12001210MG
Medchemexpress, HY-120012 10mg AZD9567 CAS:1893415-00-3 AZD9567 (compound 15) is a potent, oral active, non-steroidal and selective glucocorticoid receptor modulator (SGRM), with an IC50 of 3.8 nM. Exhibits excellent efficacy in the streptococcal cell wall (SCW) reactivation model of joint inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.