Learn More
Cayman Chemical Azelaic Acid, Purity: ≥95%, CAS: 123-99-9, 100 g
SDP
Catalog No. 502933195
Encompass_Preferred
A dicarboxylic acid with diverse biological activities; a competitive inhibitor of tyrosinase, NADH dehydrogenase, succinic dehydrogenase, and reduced ubiquitinone:cytochrome C oxidoreductase in vitro; inhibits the growth and colony formation of B16 murine as well as HMB2 and SK32 human melanoma cells; decreases DNA synthesis, induces mitochondrial damage and dilation of the RER, and reduces growth of mouse keratinocytes; bacteriostatic against various microorganisms (MICs = 0.03-0.25 M)
Catalog No. 50-293-3195 Supplier Cayman Chemical Supplier No. 23977100 G
A dicarboxylic acid with diverse biological activities; a competitive inhibitor of tyrosinase, NADH dehydrogenase, succinic dehydrogenase, and reduced ubiquitinone:cytochrome C oxidoreductase in vitro; inhibits the growth and colony formation of B16 murine as well as HMB2 and SK32 human melanoma cells; decreases DNA synthesis, induces mitochondrial damage and dilation of the RER, and reduces growth of mouse keratinocytes; bacteriostatic against various microorganisms (MICs = 0.03-0.25 M)