Learn More
Medchemexpress LLC Azelaic acid | 123-99-9 | MFCD00004432 | 98.0% | 50 G
SDP
Catalog No. 5000408149
Encompass_Preferred
Azelaic acid is a nine-carbon dicarboxylic acid. It exhibits antimicrobial activity against Propionibacterium acnes and Staphylococcus epidermidis by inhibiting microbial cellular protein synthesis. Additionally, Azelaic acid has a hypopigmentation action due to its ability to scavenge free radicals.
  • Exhibits antimicrobial activity
  • Inhibits microbial cellular protein synthesis
  • Scavenges free radicals for hypopigmentation action
  • Decreases intracellular Reactive Oxygen Species (ROS) levels
  • Increases antioxidant capacity
  • Suppresses B16, HMB2, and SK23 cell survival
  • Efficacious in treating moderate papulo-pustular rosacea
Catalog No. 50-004-08149 Supplier Medchemexpress LLC Supplier No. HYB070450G
Azelaic acid is a nine-carbon dicarboxylic acid. It exhibits antimicrobial activity against Propionibacterium acnes and Staphylococcus epidermidis by inhibiting microbial cellular protein synthesis. Additionally, Azelaic acid has a hypopigmentation action due to its ability to scavenge free radicals.
  • Exhibits antimicrobial activity
  • Inhibits microbial cellular protein synthesis
  • Scavenges free radicals for hypopigmentation action
  • Decreases intracellular Reactive Oxygen Species (ROS) levels
  • Increases antioxidant capacity
  • Suppresses B16, HMB2, and SK23 cell survival
  • Efficacious in treating moderate papulo-pustular rosacea