Learn More
Medchemexpress LLC Azelaic acid-d14 | 119176-67-9 | 99.86% | 5 MG
SDP
Catalog No. 5000408150
Encompass_Preferred
Azelaic acid-d14 is the deuterium labeled Azelaic acid. Azelaic acid is an organic compound produced by the ozonolysis of oleic acid; it is also a component of a number of hair and skin conditioners. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08150 Supplier Medchemexpress LLC Supplier No. HYB0704S5MG
Azelaic acid-d14 is the deuterium labeled Azelaic acid. Azelaic acid is an organic compound produced by the ozonolysis of oleic acid; it is also a component of a number of hair and skin conditioners. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.