Learn More
Medchemexpress LLC Azelaic acid-d14 | 119176-67-9 | 99.9% | 50 MG
SDP
Catalog No. 5000407932
Encompass_Preferred
Azelaic acid-d14 is the deuterium labeled Azelaic acid, an organic compound produced by the ozonolysis of oleic acid and a component of various hair and skin conditioners. It is intended for research use only.
  • Used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling may impact pharmacokinetic and metabolic profiles of drugs.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
Catalog No. 50-004-07932 Supplier Medchemexpress LLC Supplier No. HYB0704S50MG
Azelaic acid-d14 is the deuterium labeled Azelaic acid, an organic compound produced by the ozonolysis of oleic acid and a component of various hair and skin conditioners. It is intended for research use only.
  • Used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling may impact pharmacokinetic and metabolic profiles of drugs.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.