Learn More
AdipoGen Azelaic acid dichloride
SDP
Catalog No. 502561721
Encompass_Preferred
Chemical. CAS 123-98-8. Formula C9H14Cl2O2. MW 225.11. Azelaic acid dichloride is a bulding block for synthesis e.g. copolymers. It is the diacid chloride derivative of azelaic acid.
Catalog No. 50-256-1721 Supplier AdipoGen Supplier No. CDXA0732G025
Chemical. CAS 123-98-8. Formula C9H14Cl2O2. MW 225.11. Azelaic acid dichloride is a bulding block for synthesis e.g. copolymers. It is the diacid chloride derivative of azelaic acid.