Learn More
Cayman Chemical Azelaoyl PAF 10mg
SDP
Catalog No. 502893945
Encompass_Preferred
A degrader of cyclin K the levels of cyclin K in patient-derived CRC cells in a concentration-dependent manner inhibits cancer cell proliferation in CRC tissue samples isolated from the colon (IC50s 0.9-20.6 UM) as well as from hepatic lung and rectal metastases (IC50s 0.5-11.8 1 and 1.4-38.4 UM respectively) decreases the levels of ATM and RAD51 in patient-derived CRC tissues at 0.7 and 0.5 UM respectively reduces the number of spheroids formed and induces PARP cleavage in patient-derived CRC spheroid cultures at 3 UM
Catalog No. 50-289-3945 Supplier Cayman Chemical Supplier No. 6092410 MG
A degrader of cyclin K the levels of cyclin K in patient-derived CRC cells in a concentration-dependent manner inhibits cancer cell proliferation in CRC tissue samples isolated from the colon (IC50s 0.9-20.6 UM) as well as from hepatic lung and rectal metastases (IC50s 0.5-11.8 1 and 1.4-38.4 UM respectively) decreases the levels of ATM and RAD51 in patient-derived CRC tissues at 0.7 and 0.5 UM respectively reduces the number of spheroids formed and induces PARP cleavage in patient-derived CRC spheroid cultures at 3 UM