Learn More
Medchemexpress LLC Azelastine hydrochloride | 79307-93-0 | 99.9% | 100 MG
SDP
Catalog No. 5000408424
Encompass_Preferred
Azelastine hydrochloride is an antihistamine that functions as a potent and selective histamine 1 (H1) antagonist. It is utilized in research for various applications, including allergic rhinitis, asthma, diabetic hyperlipidemia, and SARS-CoV-2.
  • Potent and selective histamine 1 (H1) antagonist
  • Used for research in allergic rhinitis, asthma, diabetic hyperlipidemia, and SARS-CoV-2
  • Inhibits human nasal epithelial cell proliferation
  • Up-regulates H1R, M1R, and M3R levels in HNEpCs
  • Reduces blood glucose, HbA1c, serum alkaline phosphatase, and osteocalcin in diabetic hyperlipidemic rat models
  • Improves lipid profile by decreasing LDL-c and increasing HDL-c
  • Attenuates calcium deposition and aortic calcification
Catalog No. 50-004-08424 Supplier Medchemexpress LLC Supplier No. HYB0462100MG
Azelastine hydrochloride is an antihistamine that functions as a potent and selective histamine 1 (H1) antagonist. It is utilized in research for various applications, including allergic rhinitis, asthma, diabetic hyperlipidemia, and SARS-CoV-2.
  • Potent and selective histamine 1 (H1) antagonist
  • Used for research in allergic rhinitis, asthma, diabetic hyperlipidemia, and SARS-CoV-2
  • Inhibits human nasal epithelial cell proliferation
  • Up-regulates H1R, M1R, and M3R levels in HNEpCs
  • Reduces blood glucose, HbA1c, serum alkaline phosphatase, and osteocalcin in diabetic hyperlipidemic rat models
  • Improves lipid profile by decreasing LDL-c and increasing HDL-c
  • Attenuates calcium deposition and aortic calcification