Learn More
Cayman Chemical Azelastine (hydrochloride), Purity: ≥98%, CAS: 79307-93-0, 5 mg
SDP
Catalog No. 502984087
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 1.26 nM); selective for histamine H1 over histamine H3 receptors (Ki = 158.49 nM), as well as α1A- and α1B-adrenergic receptors (Ki = 50.12 nM for both); reduces nose rubbing, sneezing, and nasal mucosa expression of IL-4 in a dust mite-induced mouse model of allergic rhinitis at 0.14 µg/animal; reduces infection by a lentivirus pseudotyped with SARS-CoV-2 spike glycoprotein in HEK293 cells expressing ACE2 at 7 µg/ml
Catalog No. 50-298-4087 Supplier Cayman Chemical Supplier No. 208735 MG
A histamine H1 receptor antagonist (Ki = 1.26 nM); selective for histamine H1 over histamine H3 receptors (Ki = 158.49 nM), as well as α1A- and α1B-adrenergic receptors (Ki = 50.12 nM for both); reduces nose rubbing, sneezing, and nasal mucosa expression of IL-4 in a dust mite-induced mouse model of allergic rhinitis at 0.14 µg/animal; reduces infection by a lentivirus pseudotyped with SARS-CoV-2 spike glycoprotein in HEK293 cells expressing ACE2 at 7 µg/ml